Assay Details
| Gene Symbol: spag1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089406.2 | NP_001082875.2 | 7 - 8 | 1247 | 95 | |
| GenBank mRNA | BC139535.1 | - | 7 - 8 | 1193 | 95 | |
More Information
Set Membership: |
|

spag1a
Genomic Map