Assay Details
| Gene Symbol: epc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089409.1 | NP_001082878.1 | 3 - 4 | 512 | 77 | |
| XM_005171389.4 | XP_005171446.1 | 4 - 5 | 712 | 77 | ||
| GenBank mRNA | BC133736.1 | - | 3 - 4 | 512 | 77 | |
More Information
Set Membership: |
|

epc1b
Genomic Map