Assay Details
| Gene Symbol: slc9a6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098256.2 | NP_001091726.2 | 12 - 13 | 1692 | 68 | |
| XM_009291141.3 | XP_009289416.1 | 13 - 14 | 1837 | 68 | ||
More Information
Set Membership: |
|

slc9a6a
Genomic Map