Assay Details
| Gene Symbol: slc9a6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098256.2 | NP_001091726.2 | 15 - 16 | 1910 | 99 | |
| XM_009291141.3 | XP_009289416.1 | 16 - 17 | 2055 | 99 | ||
More Information
Set Membership: |
|

slc9a6a
Genomic Map