Assay Details
| Gene Symbol: slc9a6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098256.2 | NP_001091726.2 | 9 - 10 | 1319 | 110 | |
| XM_009291141.3 | XP_009289416.1 | 10 - 11 | 1464 | 110 | ||
More Information
Set Membership: |
|

slc9a6a
Genomic Map