Assay Details
| Gene Symbol: ccr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098743.1 | NP_001092213.1 | 2 - 3 | 94 | 71 | |
| GenBank mRNA | BC142913.1 | - | 2 - 3 | 94 | 71 | |
More Information
Set Membership: |
|

ccr7
Genomic Map