Assay Details
| Gene Symbol: plk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099245.1 | NP_001092715.1 | 4 - 5 | 578 | 68 | |
| GenBank mRNA | BC146745.1 | - | 4 - 5 | 578 | 68 | |
| HM210101.1 | - | 4 - 5 | 544 | 68 | ||
More Information
Set Membership: |
|

plk2b
Genomic Map