Assay Details
| Gene Symbol: plk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099245.1 | NP_001092715.1 | 9 - 10 | 1330 | 96 | |
| GenBank mRNA | BC146745.1 | - | 9 - 10 | 1330 | 96 | |
| HM210101.1 | - | 9 - 10 | 1296 | 96 | ||
More Information
Set Membership: |
|

plk2b
Genomic Map