Assay Details
| Gene Symbol: nrg2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099254.1 | NP_001092724.1 | 3 - 4 | 455 | 59 | |
| XM_005161327.4 | XP_005161384.1 | 3 - 4 | 771 | 59 | ||
| XM_005161328.4 | XP_005161385.1 | 3 - 4 | 772 | 59 | ||
| XM_009295459.3 | XP_009293734.1 | 3 - 4 | 772 | 59 | ||
| XM_009295460.3 | XP_009293735.1 | 3 - 4 | 772 | 59 | ||
| XM_009295461.3 | XP_009293736.1 | 3 - 4 | 536 | 59 | ||
| XM_009295462.3 | XP_009293737.1 | 3 - 4 | 700 | 59 | ||
| GenBank mRNA | BC139893.1 | - | 3 - 4 | 455 | 59 | |
| FJ593490.1 | - | 3 - 4 | 455 | 59 | ||
| FJ593491.1 | - | 3 - 4 | 455 | 59 | ||
| FJ593492.1 | - | 3 - 4 | 455 | 59 | ||
| FJ593493.1 | - | 3 - 4 | 455 | 59 | ||
More Information
Set Membership: |
|

nrg2a
Genomic Map