Assay Details
| Gene Symbol: nrg2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099254.1 | NP_001092724.1 | 5 - 6 | 666 | 72 | |
| XM_005161327.4 | XP_005161384.1 | 5 - 6 | 982 | 72 | ||
| XM_009295460.3 | XP_009293735.1 | 5 - 6 | 983 | 72 | ||
| GenBank mRNA | BC139893.1 | - | 5 - 6 | 666 | 72 | |
| FJ593490.1 | - | 5 - 6 | 666 | 72 | ||
| FJ593493.1 | - | 5 - 6 | 666 | 72 | ||
More Information
Set Membership: |
|

nrg2a
Genomic Map