Assay Details
| Gene Symbol: ptbp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100007.1 | NP_001093477.1 | 14 - 15 | 1589 | 71 | |
| XM_005163345.4 | XP_005163402.1 | 13 - 14 | 1571 | 71 | ||
More Information
Set Membership: |
|

ptbp2b
Genomic Map