Assay Details
| Gene Symbol: msrb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105128.3 | NP_001098598.2 | 2 - 3 | 319 | 103 | |
| GenBank mRNA | BC081680.1 | - | 2 - 3 | 270 | 103 | |
| BC152619.1 | - | 2 - 3 | 287 | 103 | ||
| EH578474.1 | - | 2 - 3 | 341 | 103 | ||
More Information
Set Membership: |
|

msrb1b
Genomic Map