Assay Details
| Gene Symbol: sh3gl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105606.1 | NP_001099076.1 | 1 - 2 | 60 | 65 | |
| NM_200301.1 | NP_956595.1 | 1 - 2 | 178 | 65 | ||
| XM_005168418.1 | XP_005168475.1 | 1 - 2 | 216 | 65 | ||
| GenBank mRNA | BC049513.1 | - | 1 - 2 | 178 | 65 | |
| BC153638.1 | - | 1 - 2 | 60 | 65 | ||
More Information
Set Membership: |
|

sh3gl1a
Genomic Map