Assay Details
| Gene Symbol: cpsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131009.1 | NP_571084.1 | 4 - 5 | 693 | 66 | |
| XM_021469299.1 | XP_021324974.1 | 4 - 5 | 728 | 66 | ||
| GenBank mRNA | BC045289.1 | - | 4 - 5 | 696 | 66 | |
| BC164456.1 | - | 4 - 5 | 434 | 66 | ||
| U70479.1 | - | 4 - 5 | 693 | 66 | ||
More Information
Set Membership: |
|

cpsf4
Genomic Map