Assay Details
| Gene Symbol: wt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131046.2 | NP_571121.1 | 5 - 6 | 1127 | 60 | |
| GenBank mRNA | AF144550.1 | - | 5 - 6 | 876 | 60 | |
| BC162349.1 | - | 5 - 6 | 836 | 60 | ||
| BC162638.1 | - | 5 - 6 | 836 | 60 | ||
More Information
Set Membership: |
|

wt1a
Genomic Map