Assay Details
| Gene Symbol: psmb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131151.1 | NP_571226.1 | 1 - 2 | 232 | 148 | |
| GenBank mRNA | AF032391.1 | - | 1 - 2 | 181 | 148 | |
| AF155578.1 | - | 1 - 2 | 181 | 148 | ||
| BC071478.1 | - | 1 - 2 | 232 | 148 | ||
| BC153589.1 | - | 1 - 2 | 233 | 148 | ||
More Information
Set Membership: |
|

psmb5
Genomic Map