Assay Details
Gene Symbol: ascl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131219.1 | NP_571294.1 | 1 - 2 | 721 | 69 | |
GenBank mRNA | BC098521.1 | - | 1 - 2 | 732 | 69 | |
U14587.1 | - | 1 - 2 | 721 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
