Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 12 - 13 | 1451 | 72 | |
| GenBank mRNA | AF130459.1 | - | 12 - 13 | 1452 | 72 | |
| BC115197.1 | - | 1 - 2 | 75 | 72 | ||
| BC134876.1 | - | 1 - 2 | 91 | 72 | ||
More Information
Set Membership: |
|

trpc4apa
Genomic Map