Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 13 - 14 | 1527 | 130 | |
| GenBank mRNA | AF130459.1 | - | 13 - 14 | 1528 | 130 | |
| BC115197.1 | - | 2 - 3 | 151 | 130 | ||
| BC134876.1 | - | 2 - 3 | 167 | 130 | ||
More Information
Set Membership: |
|

trpc4apa
Genomic Map