Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 14 - 15 | 1628 | 82 | |
| GenBank mRNA | AF130459.1 | - | 14 - 15 | 1629 | 82 | |
| BC099990.1 | - | 3 - 4 | 238 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC099990.1 transcripts.
Set Membership: |
|

trpc4apa
Genomic Map