Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 2 - 3 | 265 | 151 | |
| GenBank mRNA | AF130459.1 | - | 2 - 3 | 266 | 151 | |
More Information
Set Membership: |
|

trpc4apa
Genomic Map