Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 3 - 4 | 385 | 94 | |
| GenBank mRNA | AF130459.1 | - | 3 - 4 | 386 | 94 | |
More Information
Set Membership: |
|

trpc4apa
Genomic Map