Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 4 - 5 | 430 | 69 | |
| GenBank mRNA | AF130459.1 | - | 4 - 5 | 431 | 69 | |
More Information
Set Membership: |
|

trpc4apa
Genomic Map