Assay Details
Gene Symbol: trpc4apa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131569.2 | NP_571644.1 | 7 - 8 | 823 | 96 | |
GenBank mRNA | AF130459.1 | - | 7 - 8 | 824 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
