Assay Details
| Gene Symbol: trpc4apa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131569.2 | NP_571644.1 | 9 - 10 | 1154 | 68 | |
| GenBank mRNA | AF130459.1 | - | 9 - 10 | 1155 | 68 | |
More Information
Set Membership: |
|

trpc4apa
Genomic Map