Assay Details
| Gene Symbol: pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131612.2 | NP_571687.1 | 4 - 5 | 853 | 74 | |
| XM_017351360.2 | XP_017206849.1 | 4 - 5 | 844 | 74 | ||
| GenBank mRNA | AJ245965.1 | - | 776 | 74 | ||
| BC162623.1 | - | 4 - 5 | 853 | 74 | ||
More Information
Set Membership: |
|

pbx2
Genomic Map