Assay Details
| Gene Symbol: pbx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131615.1 | NP_571690.1 | 6 - 7 | 1358 | 111 | |
| GenBank mRNA | AJ245963.1 | - | 6 - 7 | 1358 | 111 | |
More Information
Set Membership: |
|

pbx1a
Genomic Map