Assay Details
| Gene Symbol: jag1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131863.2 | NP_571938.2 | 21 - 22 | 2823 | 88 | |
| GenBank mRNA | AF229451.1 | - | 21 - 22 | 2785 | 88 | |
| AY221106.1 | - | 21 - 22 | 2824 | 88 | ||
More Information
Set Membership: |
|

jag1b
Genomic Map