Assay Details
| Gene Symbol: jag1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131863.2 | NP_571938.2 | 4 - 5 | 952 | 54 | |
| GenBank mRNA | AF229451.1 | - | 4 - 5 | 914 | 54 | |
| AY221106.1 | - | 4 - 5 | 953 | 54 | ||
More Information
Set Membership: |
|

jag1b
Genomic Map