Assay Details
| Gene Symbol: ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152949.1 | NP_694481.1 | 3 - 4 | 699 | 75 | |
| XM_021480961.1 | XP_021336636.1 | 2 - 3 | 807 | 75 | ||
| GenBank mRNA | AF234784.2 | - | 3 - 4 | 699 | 75 | |
| BC045840.1 | - | 3 - 4 | 699 | 75 | ||
| BC068323.1 | - | 3 - 4 | 514 | 75 | ||
| BC151890.1 | - | 3 - 4 | 694 | 75 | ||
More Information
Set Membership: |
|

ccna2
Genomic Map