Assay Details
| Gene Symbol: id3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152967.1 | NP_694499.1 | 2 - 3 | 509 | 98 | |
| GenBank mRNA | AY065841.1 | - | 2 - 3 | 509 | 98 | |
| BC066743.1 | - | 2 - 3 | 505 | 98 | ||
| BC152276.1 | - | 2 - 3 | 525 | 98 | ||
More Information
Set Membership: |
|

id3
Genomic Map