Assay Details
| Gene Symbol: ckbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173222.1 | NP_775329.1 | 6 - 7 | 859 | 111 | |
| GenBank mRNA | AY055849.1 | - | 6 - 7 | 859 | 111 | |
| BC049529.1 | - | 6 - 7 | 875 | 111 | ||
| BC064285.1 | - | 6 - 7 | 863 | 111 | ||
More Information
Set Membership: |
|

ckbb
Genomic Map