Assay Details
| Gene Symbol: taf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173260.1 | NP_775367.1 | 5 - 6 | 436 | 129 | |
| XM_005173300.4 | XP_005173357.1 | 5 - 6 | 600 | 129 | ||
| GenBank mRNA | AF506232.1 | - | 5 - 6 | 436 | 129 | |
| BC071324.1 | - | 5 - 6 | 400 | 129 | ||
| BC154771.1 | - | 5 - 6 | 570 | 129 | ||
More Information
Set Membership: |
|

taf7
Genomic Map