Assay Details
| Gene Symbol: taf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173260.1 | NP_775367.1 | 9 - 10 | 795 | 108 | |
| XM_005173300.4 | XP_005173357.1 | 9 - 10 | 959 | 108 | ||
| GenBank mRNA | AF506232.1 | - | 9 - 10 | 795 | 108 | |
| BC071324.1 | - | 9 - 10 | 759 | 108 | ||
| BC154771.1 | - | 9 - 10 | 929 | 108 | ||
More Information
Set Membership: |
|

taf7
Genomic Map