Assay Details
| Gene Symbol: anxa5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181757.2 | NP_861422.2 | 3 - 4 | 198 | 79 | |
| GenBank mRNA | AY178799.1 | - | 3 - 4 | 174 | 79 | |
| BC046873.1 | - | 3 - 4 | 198 | 79 | ||
| BC065430.1 | - | 3 - 4 | 156 | 79 | ||
| BC164112.1 | - | 1 - 2 | 122 | 79 | ||
More Information
Set Membership: |
|

anxa5b
Genomic Map