Assay Details
| Gene Symbol: anxa5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181757.2 | NP_861422.2 | 4 - 5 | 287 | 72 | |
| GenBank mRNA | AY178799.1 | - | 4 - 5 | 263 | 72 | |
| BC046873.1 | - | 4 - 5 | 287 | 72 | ||
| BC065430.1 | - | 4 - 5 | 245 | 72 | ||
| BC155280.1 | - | 4 - 5 | 287 | 72 | ||
| BC164112.1 | - | 2 - 3 | 211 | 72 | ||
More Information
Set Membership: |
|

anxa5b
Genomic Map