Assay Details
| Gene Symbol: anxa5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181757.2 | NP_861422.2 | 6 - 7 | 498 | 112 | |
| GenBank mRNA | AY178799.1 | - | 6 - 7 | 474 | 112 | |
| BC046873.1 | - | 6 - 7 | 498 | 112 | ||
| BC065430.1 | - | 6 - 7 | 456 | 112 | ||
| BC155280.1 | - | 6 - 7 | 498 | 112 | ||
| BC164112.1 | - | 4 - 5 | 422 | 112 | ||
More Information
Set Membership: |
|

anxa5b
Genomic Map