Assay Details
| Gene Symbol: elf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199219.2 | NP_954689.1 | 1 - 2 | 143 | 66 | |
| GenBank mRNA | BC056554.1 | - | 1 - 2 | 143 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

elf2b| Gene Symbol: elf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199219.2 | NP_954689.1 | 1 - 2 | 143 | 66 | |
| GenBank mRNA | BC056554.1 | - | 1 - 2 | 143 | 66 | |
Set Membership: |
|