Assay Details
| Gene Symbol: elf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199219.2 | NP_954689.1 | 2 - 3 | 243 | 65 | |
| XM_005159828.2 | XP_005159885.1 | 4 - 5 | 782 | 65 | ||
| XM_005159829.4 | XP_005159886.1 | 4 - 5 | 812 | 65 | ||
| XM_005159830.4 | XP_005159887.1 | 4 - 5 | 539 | 65 | ||
| GenBank mRNA | BC056554.1 | - | 2 - 3 | 243 | 65 | |
| U84616.1 | - | 4 - 5 | 972 | 65 | ||
More Information
Set Membership: |
|

elf2b
Genomic Map