Assay Details
| Gene Symbol: elf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199219.2 | NP_954689.1 | 4 - 5 | 505 | 108 | |
| XM_005159828.2 | XP_005159885.1 | 6 - 7 | 1044 | 108 | ||
| XM_005159829.4 | XP_005159886.1 | 6 - 7 | 1074 | 108 | ||
| XM_005159830.4 | XP_005159887.1 | 6 - 7 | 801 | 108 | ||
| GenBank mRNA | BC056554.1 | - | 4 - 5 | 505 | 108 | |
| U84616.1 | - | 6 - 7 | 1234 | 108 | ||
More Information
Set Membership: |
|

elf2b
Genomic Map