Assay Details
| Gene Symbol: dtnba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324537.1 | NP_001311466.1 | 8 - 9 | 1109 | 79 | |
| NM_199709.2 | NP_956003.2 | 7 - 8 | 1024 | 79 | ||
| XM_005168526.4 | XP_005168583.1 | 9 - 10 | 1154 | 79 | ||
| XM_005168527.4 | XP_005168584.1 | 9 - 10 | 1154 | 79 | ||
| XM_005168528.4 | XP_005168585.1 | 9 - 10 | 1154 | 79 | ||
| XM_009294981.3 | XP_009293256.1 | 8 - 9 | 1111 | 79 | ||
| GenBank mRNA | AY423026.1 | - | 7 - 8 | 79 | ||
| DQ516346.1 | - | 8 - 9 | 79 | |||
More Information
Set Membership: |
|

dtnba
Genomic Map