Assay Details
| Gene Symbol: chp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199715.1 | NP_956009.1 | 3 - 4 | 381 | 122 | |
| GenBank mRNA | AY398327.1 | - | 3 - 4 | 454 | 122 | |
| BC054566.1 | - | 3 - 4 | 381 | 122 | ||
More Information
Set Membership: |
|

chp1
Genomic Map