Assay Details
| Gene Symbol: dgat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199730.1 | NP_956024.1 | 15 - 16 | 1342 | 70 | |
| GenBank mRNA | BC063970.1 | - | 15 - 16 | 1342 | 70 | |
More Information
Set Membership: |
|

dgat1a
Genomic Map