Assay Details
| Gene Symbol: alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199867.1 | NP_956161.1 | 8 - 9 | 985 | 83 | |
| GenBank mRNA | BC055554.1 | - | 8 - 9 | 985 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

alg1| Gene Symbol: alg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199867.1 | NP_956161.1 | 8 - 9 | 985 | 83 | |
| GenBank mRNA | BC055554.1 | - | 8 - 9 | 985 | 83 | |
Set Membership: |
|