Assay Details
| Gene Symbol: nme2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199970.2 | 4 - 5 | 427 | 68 | ||
| XM_021470954.1 | XP_021326629.1 | 4 - 5 | 509 | 68 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nme2a| Gene Symbol: nme2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199970.2 | 4 - 5 | 427 | 68 | ||
| XM_021470954.1 | XP_021326629.1 | 4 - 5 | 509 | 68 | ||
Set Membership: |
|