Assay Details
| Gene Symbol: rps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200026.1 | NP_956320.1 | 4 - 5 | 406 | 64 | |
| GenBank mRNA | BC059561.1 | - | 4 - 5 | 406 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rps14| Gene Symbol: rps14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200026.1 | NP_956320.1 | 4 - 5 | 406 | 64 | |
| GenBank mRNA | BC059561.1 | - | 4 - 5 | 406 | 64 | |
Set Membership: |
|