Assay Details
| Gene Symbol: rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200027.1 | NP_956321.1 | 4 - 5 | 354 | 143 | |
| XM_017353523.1 | XP_017209012.1 | 5 - 6 | 368 | 143 | ||
| GenBank mRNA | AY763500.1 | - | 4 - 5 | 356 | 143 | |
| BC045950.1 | - | 4 - 5 | 354 | 143 | ||
| BC164313.1 | - | 4 - 5 | 365 | 143 | ||
More Information
Set Membership: |
|

rpl10
Genomic Map