Assay Details
| Gene Symbol: ythdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200250.1 | NP_956544.1 | 3 - 4 | 271 | 63 | |
| XM_005158763.4 | XP_005158820.1 | 3 - 4 | 300 | 63 | ||
| GenBank mRNA | BC047846.1 | - | 3 - 4 | 271 | 63 | |
More Information
Set Membership: |
|

ythdf2
Genomic Map