Assay Details
| Gene Symbol: taf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200286.1 | NP_956580.1 | 2 - 3 | 277 | 62 | |
| XM_005162303.4 | XP_005162360.1 | 2 - 3 | 367 | 62 | ||
| GenBank mRNA | BC049422.1 | - | 2 - 3 | 277 | 62 | |
| BC065457.1 | - | 2 - 3 | 314 | 62 | ||
More Information
Set Membership: |
|

taf8
Genomic Map