Assay Details
| Gene Symbol: zgc:56596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200300.1 | NP_956594.1 | 2 - 3 | 277 | 95 | |
| GenBank mRNA | BC049498.1 | - | 2 - 3 | 277 | 95 | |
More Information
Set Membership: |
|

zgc:56596
Genomic Map